siRNA Details
Gene Symbol: Cldn19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008514.1 | NP_001008514.1 | 3 | 484 | ||
XM_006238756.2 | XP_006238818.1 | 3 | 956 | |||
XM_008763997.1 | XP_008762219.1 | 4 | 941 | |||
GenBank mRNA | AF497645.1 | - | 3 | 400 | ||
BC079172.1 | - | 3 | 484 | 7129992 |
