siRNA Details
Gene Symbol: Grhl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106690.1 | NP_001100160.1 | 2 | 256 | ||
XM_006239149.2 | XP_006239211.1 | 2 | 398 | |||
XM_008764216.1 | XP_008762438.1 | 2 | 398 |
