siRNA Details
Gene Symbol: Sdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100539.1 | NP_001094009.1 | 3 | 245 | ||
GenBank mRNA | BC158620.1 | - | 3 | 245 | 7446362 | |
DQ403001.1 | - | 2 | 150 | |||
FQ215046.1 | - | 3 | 241 |
