siRNA Details
Gene Symbol: Nmnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037556.2 | NP_001032645.2 | 4, 5 | 546 | ||
XM_006239429.2 | XP_006239491.1 | 4, 5 | 1180 | |||
XM_006239430.2 | XP_006239492.1 | 4, 5 | 765 | |||
XM_006239431.2 | XP_006239493.1 | 3, 4 | 507 | |||
XM_008764237.1 | XP_008762459.1 | 5, 6 | 703 | |||
XM_008764238.1 | XP_008762460.1 | 4, 5 | 528 | |||
GenBank mRNA | AY788917.1 | - | 4, 5 | 540 | ||
BC127446.1 | - | 4, 5 | 546 | 7383226 |
