siRNA Details
Gene Symbol: Cpsf3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033892.1 | NP_001029064.1 | 4, 5 | 510 | ||
XM_006239549.2 | XP_006239611.1 | 4, 5 | 552 | |||
XM_006239550.2 | XP_006239612.1 | 4, 5 | 552 | |||
XM_006239551.2 | XP_006239613.1 | 4, 5 | 552 | |||
XM_006239552.2 | XP_006239614.1 | 4, 5 | 580 | |||
GenBank mRNA | BC105303.1 | - | 4, 5 | 510 | 7110348 |
