siRNA Details
Gene Symbol: Ylpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271258.1 | NP_001258187.1 | 16 | 6087 | ||
GenBank mRNA | FQ219197.1 | - | 3 | 200 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ylpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271258.1 | NP_001258187.1 | 16 | 6087 | ||
GenBank mRNA | FQ219197.1 | - | 3 | 200 |