siRNA Details
Gene Symbol: Ift43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134525.1 | NP_001127997.1 | 3 | 247 | ||
XM_006240370.2 | XP_006240432.1 | 3 | 233 | |||
XM_006240372.2 | XP_006240434.1 | 3 | 233 |
