siRNA Details
Gene Symbol: Tdrd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764983.1 | XP_008763205.1 | 26 | 3017 | ||
XM_008764984.1 | XP_008763206.1 | 26 | 3017 | |||
XM_008764985.1 | XP_008763207.1 | 26 | 2984 | |||
XM_008764986.1 | XP_008763208.1 | 26 | 2984 | |||
XM_008764987.1 | XP_008763209.1 | 25 | 2900 | |||
XM_008764988.1 | XP_008763210.1 | 26 | 2788 | |||
GenBank mRNA | BC087164.1 | - | 17 | 3162 | ||
BC104675.1 | - | 3 | 978 |
