siRNA Details
Gene Symbol: Tdrd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764983.1 | XP_008763205.1 | 9, 10 | 1292 | ||
XM_008764984.1 | XP_008763206.1 | 9, 10 | 1292 | |||
XM_008764985.1 | XP_008763207.1 | 9, 10 | 1292 | |||
XM_008764986.1 | XP_008763208.1 | 9, 10 | 1292 | |||
XM_008764987.1 | XP_008763209.1 | 9, 10 | 1292 | |||
XM_008764988.1 | XP_008763210.1 | 9, 10 | 1063 | |||
GenBank mRNA | BC087164.1 | - | 2, 3 | 184 |
