siRNA Details
Gene Symbol: Morc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106764.1 | NP_001100234.1 | 2 | 531 |
Gene Symbol: LOC100911941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752187.3 | XP_003752235.2 | 1 | 334 |
