siRNA Details
Gene Symbol: Mknk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011985.1 | NP_001011985.1 | 5, 6 | 483 | ||
XM_006240883.2 | XP_006240945.1 | 5, 6 | 572 | |||
XM_006240884.1 | XP_006240946.1 | 4, 5 | 460 | |||
GenBank mRNA | BC085941.1 | - | 5, 6 | 483 | 7124839 | |
FQ211808.1 | - | 5, 6 | 526 |
