siRNA Details
Gene Symbol: LOC100362054 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758861.1 | XP_008757083.1 | 4, 5 | 1339 |
Gene Symbol: Zfp494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_234948.3 | XP_234948.1 | 1, 2 | 404 |
