siRNA Details
Gene Symbol: Ccdc53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106776.1 | NP_001100246.1 | 3 | 315 | ||
XM_006241199.2 | XP_006241261.1 | 3 | 287 | |||
GenBank mRNA | FQ221863.1 | - | 2 | 90 |
