siRNA Details
Gene Symbol: Mars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127659.1 | NP_001121131.1 | 3, 4 | 300 | ||
XM_006241451.2 | XP_006241513.1 | 3, 4 | 413 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127659.1 | NP_001121131.1 | 3, 4 | 300 | ||
XM_006241451.2 | XP_006241513.1 | 3, 4 | 413 |