siRNA Details
Gene Symbol: Ttll12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135922.1 | NP_001129394.1 | 6, 7 | 925 | ||
XM_008765684.1 | XP_008763906.1 | 6, 7 | 1059 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ttll12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135922.1 | NP_001129394.1 | 6, 7 | 925 | ||
XM_008765684.1 | XP_008763906.1 | 6, 7 | 1059 |