siRNA Details
Gene Symbol: Gxylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100887.1 | NP_001094357.1 | 8 | 1225 | ||
XM_006242192.2 | XP_006242254.1 | 7 | 1133 | |||
GenBank mRNA | AY623036.1 | - | 6 | 828 | ||
BC167068.1 | - | 7 | 979 | 8363586 |
