siRNA Details
Gene Symbol: Kansl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004244.2 | NP_001004244.2 | 4 | 628 | ||
XM_008765691.1 | XP_008763913.1 | 5 | 956 | |||
XM_008765692.1 | XP_008763914.1 | 5 | 712 | |||
XM_008765693.1 | XP_008763915.1 | 5 | 712 | |||
XM_008765694.1 | XP_008763916.1 | 3 | 528 | |||
GenBank mRNA | BC079169.1 | - | 3 | 510 | 7129861 | |
BC169120.1 | - | 4 | 871 | |||
CK474940.1 | - | 4 | 571 | |||
CO404725.1 | - | 3 | 528 | |||
EF076771.1 | - | 4 | 629 |
Gene Symbol: LOC103690317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765853.1 | XP_008764075.1 | 956 | |||
XM_008765854.1 | XP_008764076.1 | 712 | ||||
XM_008765855.1 | XP_008764077.1 | 712 | ||||
XM_008765856.1 | XP_008764078.1 | 916 | ||||
XM_008765857.1 | XP_008764079.1 | 528 |
