siRNA Details
Gene Symbol: Alg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013951.1 | NP_001013973.1 | 4 | 413 | ||
GenBank mRNA | BC088233.1 | - | 4 | 413 | 7324702 | |
FQ216283.1 | - | 4 | 425 | |||
FQ229089.1 | - | 4 | 439 |
