siRNA Details
Gene Symbol: Ccdc153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013953.1 | NP_001013975.1 | 5 | 727 | ||
XM_006242913.2 | XP_006242975.1 | 6 | 1209 | |||
GenBank mRNA | BC089910.1 | - | 5 | 727 | 7304742 |
