siRNA Details
Gene Symbol: Bbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106826.1 | NP_001100296.1 | 12 | 1022 | ||
XM_008766224.1 | XP_008764446.1 | 11 | 1354 | |||
XM_008766225.1 | XP_008764447.1 | 7 | 1165 | |||
XM_008766226.1 | XP_008764448.1 | 6 | 821 |
