siRNA Details
Gene Symbol: Celf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106827.1 | NP_001100297.1 | 4 | 409 | ||
XM_008766227.1 | XP_008764449.1 | 4 | 653 | |||
XM_008766228.1 | XP_008764450.1 | 4 | 653 | |||
XM_008766229.1 | XP_008764451.1 | 4 | 653 | |||
XM_008766230.1 | XP_008764452.1 | 4 | 653 |
