siRNA Details
Gene Symbol: Parp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106828.1 | NP_001100298.1 | 19, 20 | 1965 | ||
XM_006243191.1 | XP_006243253.1 | 19, 20 | 2249 | |||
XM_006243192.2 | XP_006243254.1 | 18, 19 | 2907 | |||
XM_006243193.2 | XP_006243255.1 | 18, 19 | 1776 | |||
XM_008766231.1 | XP_008764453.1 | 18, 19 | 2904 | |||
XM_008766232.1 | XP_008764454.1 | 18, 19 | 2847 | |||
XM_008766233.1 | XP_008764455.1 | 18, 19 | 2844 |
