siRNA Details
Gene Symbol: Morf4l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011999.1 | NP_001011999.1 | 12 | 1058 | ||
XM_008766445.1 | XP_008764667.1 | 13 | 1141 | |||
GenBank mRNA | BC078910.1 | - | 12 | 1058 | 7131534 | |
FQ213551.1 | - | 6 | 578 |
