siRNA Details
Gene Symbol: Tfdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106847.2 | NP_001100317.2 | 9 | 960 | ||
XM_006243556.2 | XP_006243618.1 | 1264 | ||||
XM_006243558.2 | XP_006243620.1 | 9 | 2363 | |||
XM_006243561.2 | XP_006243623.1 | 8 | 2323 | |||
XM_006243563.1 | XP_006243625.1 | 11 | 1372 | |||
XM_006243566.2 | XP_006243628.1 | 1565 | ||||
XM_006243567.2 | XP_006243629.1 | 10 | 2687 | |||
XM_006243568.2 | XP_006243630.1 | 11 | 2784 | |||
XM_006243569.2 | XP_006243631.1 | 10 | 1498 | |||
XM_006243571.2 | XP_006243633.1 | 10 | 1451 | |||
XM_006243572.2 | XP_006243634.1 | 8 | 2319 | |||
XM_008766479.1 | XP_008764701.1 | 10 | 942 | |||
XM_008766480.1 | XP_008764702.1 | 10 | 938 | |||
XM_008766481.1 | XP_008764703.1 | 10 | 942 | |||
XM_008766482.1 | XP_008764704.1 | 14 | 1479 | |||
XM_008766483.1 | XP_008764705.1 | 10 | 1265 | |||
XM_008766484.1 | XP_008764706.1 | 14 | 1333 | |||
XM_008766485.1 | XP_008764707.1 | 13 | 1237 | |||
XM_008766487.1 | XP_008764709.1 | 11 | 1068 | |||
XM_008766488.1 | XP_008764710.1 | 8 | 778 | |||
GenBank mRNA | BC168961.1 | - | 9 | 960 | 7444676 |
