siRNA Details
Gene Symbol: Tfdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106847.2 | NP_001100317.2 | 9, 10 | 1020 | ||
XM_006243556.2 | XP_006243618.1 | 1324 | ||||
XM_006243558.2 | XP_006243620.1 | 9, 10 | 2423 | |||
XM_006243561.2 | XP_006243623.1 | 8, 9 | 2383 | |||
XM_006243563.1 | XP_006243625.1 | 11, 12 | 1432 | |||
XM_006243566.2 | XP_006243628.1 | 1625 | ||||
XM_006243567.2 | XP_006243629.1 | 10, 11 | 2747 | |||
XM_006243568.2 | XP_006243630.1 | 11, 12 | 2844 | |||
XM_006243569.2 | XP_006243631.1 | 10, 11 | 1558 | |||
XM_006243571.2 | XP_006243633.1 | 10, 11 | 1511 | |||
XM_006243572.2 | XP_006243634.1 | 8, 9 | 2379 | |||
XM_008766479.1 | XP_008764701.1 | 10, 11 | 1002 | |||
XM_008766480.1 | XP_008764702.1 | 10, 11 | 998 | |||
XM_008766481.1 | XP_008764703.1 | 10, 11 | 1002 | |||
XM_008766482.1 | XP_008764704.1 | 14, 15 | 1539 | |||
XM_008766483.1 | XP_008764705.1 | 10, 11 | 1325 | |||
XM_008766484.1 | XP_008764706.1 | 14, 15 | 1393 | |||
XM_008766485.1 | XP_008764707.1 | 13, 14 | 1297 | |||
XM_008766487.1 | XP_008764709.1 | 11, 12 | 1128 | |||
XM_008766488.1 | XP_008764710.1 | 8, 9 | 838 | |||
GenBank mRNA | BC168961.1 | - | 9, 10 | 1020 | 7444676 |
