siRNA Details
Gene Symbol: Tfdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106847.2 | NP_001100317.2 | 11 | 1238 | ||
XM_006243556.2 | XP_006243618.1 | 1542 | ||||
XM_006243558.2 | XP_006243620.1 | 11 | 2641 | |||
XM_006243561.2 | XP_006243623.1 | 10 | 2601 | |||
XM_006243563.1 | XP_006243625.1 | 13 | 1650 | |||
XM_006243566.2 | XP_006243628.1 | 1843 | ||||
XM_006243567.2 | XP_006243629.1 | 12 | 2965 | |||
XM_006243568.2 | XP_006243630.1 | 13 | 3062 | |||
XM_006243569.2 | XP_006243631.1 | 12 | 1776 | |||
XM_006243571.2 | XP_006243633.1 | 12 | 1729 | |||
XM_006243572.2 | XP_006243634.1 | 10 | 2597 | |||
XM_008766479.1 | XP_008764701.1 | 12 | 1220 | |||
XM_008766480.1 | XP_008764702.1 | 12 | 1216 | |||
XM_008766481.1 | XP_008764703.1 | 12 | 1220 | |||
XM_008766482.1 | XP_008764704.1 | 16 | 1757 | |||
XM_008766483.1 | XP_008764705.1 | 12 | 1543 | |||
XM_008766484.1 | XP_008764706.1 | 16 | 1611 | |||
XM_008766485.1 | XP_008764707.1 | 15 | 1515 | |||
XM_008766487.1 | XP_008764709.1 | 13 | 1346 | |||
XM_008766488.1 | XP_008764710.1 | 10 | 1056 | |||
GenBank mRNA | BC168961.1 | - | 11 | 1238 | 7444676 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178667, NM_001184706, NM_001184710, NM_001184711, NM_001184708, and others.
Gene Ontology Categories:
