siRNA Details
Gene Symbol: Scap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100966.1 | NP_001094436.1 | 11 | 1466 | ||
XM_006243922.2 | XP_006243984.1 | 10 | 1411 | |||
XM_006243923.2 | XP_006243985.1 | 11 | 1482 | |||
XM_006243924.2 | XP_006243986.1 | 10 | 3362 | |||
XM_006243925.2 | XP_006243987.1 | 10 | 1303 | |||
GenBank mRNA | BC131852.1 | - | 11 | 1466 | 7189794 |
