siRNA Details
Gene Symbol: Susd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766656.1 | XP_008764878.1 | 4 | 1595 | ||
XM_236674.7 | XP_236674.5 | 5 | 1784 |

More Information
Other Species Targeted

Gene Ontology Categories:
