siRNA Details
Gene Symbol: Slc25a41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106874.1 | NP_001100344.1 | 7 | 947 | ||
XM_006244305.1 | XP_006244367.1 | 7 | 996 | |||
XM_008766754.1 | XP_008764976.1 | 7 | 1057 | |||
GenBank mRNA | FM165287.1 | - | 7 | 884 |
