siRNA Details
Gene Symbol: Mllt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106876.1 | NP_001100346.1 | 4 | 492 | ||
XM_006244307.2 | XP_006244369.1 | 4 | 488 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_022328, BC053005, AK144605, AF312858, AF298887, and others.
Gene Ontology Categories:
