siRNA Details
Gene Symbol: Enpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106892.1 | NP_001100362.1 | 2 | 706 | ||
XM_006244598.2 | XP_006244660.1 | 3 | 2102 | |||
XM_006244600.2 | XP_006244662.1 | 2 | 1987 | |||
XM_006244601.2 | XP_006244663.1 | 4 | 1751 | |||
XM_006244602.2 | XP_006244664.1 | 2 | 688 | |||
XM_008766852.1 | XP_008765074.1 | 3 | 802 | |||
XM_008766853.1 | XP_008765075.1 | 2 | 596 |
