siRNA Details
Gene Symbol: Ankrd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191807.3 | NP_001178736.3 | 642 | |||
GenBank mRNA | FQ158391.1 | - | 642 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ankrd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191807.3 | NP_001178736.3 | 642 | |||
GenBank mRNA | FQ158391.1 | - | 642 |