siRNA Details
Gene Symbol: Rnf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012004.2 | NP_001012004.2 | 10 | 903 | ||
XM_006245172.2 | XP_006245234.1 | 10 | 931 | |||
XM_006245173.1 | XP_006245235.1 | 10 | 1784 | |||
GenBank mRNA | BM382839.1 | - | 1 | 556 | ||
CB775437.1 | - | 3 | 276 | |||
CK845002.1 | - | 1 | 560 |
