siRNA Details
Gene Symbol: Col4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008332.1 | NP_001008333.1 | 4 | 325 | ||
GenBank mRNA | BC086403.1 | - | 4 | 325 | 7304199 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Col4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008332.1 | NP_001008333.1 | 4 | 325 | ||
GenBank mRNA | BC086403.1 | - | 4 | 325 | 7304199 |