siRNA Details
Gene Symbol: Fam174a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012350.1 | NP_001012350.1 | 1 | 473 | ||
XM_006245579.2 | XP_006245641.1 | 1 | 599 | |||
GenBank mRNA | BC089835.1 | - | 1 | 473 | 7326083 |
