siRNA Details
Gene Symbol: Efcab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106930.1 | NP_001100400.1 | 2 | 263 | ||
XM_008768871.1 | XP_008767093.1 | 2 | 294 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Efcab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106930.1 | NP_001100400.1 | 2 | 263 | ||
XM_008768871.1 | XP_008767093.1 | 2 | 294 |