siRNA Details
Gene Symbol: Eda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257110.2 | XP_006257172.1 | 1 | 375 | ||
XM_006257111.2 | XP_006257173.1 | 1 | 375 | |||
XM_006257112.2 | XP_006257174.1 | 1 | 375 | |||
XM_008773301.1 | XP_008771523.1 | 1 | 375 | |||
XM_228582.8 | XP_228582.6 | 1 | 375 |

More Information
Other Species Targeted

Gene Ontology Categories:
