siRNA Details
Gene Symbol: Adcy9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767525.1 | XP_008765747.1 | 2 | 1100 | ||
XM_008767526.1 | XP_008765748.1 | 2 | 1100 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adcy9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767525.1 | XP_008765747.1 | 2 | 1100 | ||
XM_008767526.1 | XP_008765748.1 | 2 | 1100 |