siRNA Details
Gene Symbol: Abca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246039.2 | XP_006246101.1 | 5 | 810 | ||
XM_220219.9 | XP_220219.6 | 4 | 669 | |||
GenBank mRNA | BC088202.1 | - | 3 | 455 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Abca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246039.2 | XP_006246101.1 | 5 | 810 | ||
XM_220219.9 | XP_220219.6 | 4 | 669 | |||
GenBank mRNA | BC088202.1 | - | 3 | 455 |