siRNA Details
Gene Symbol: Guk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013115.2 | NP_001013133.2 | 5 | 434 | ||
XM_006246477.2 | XP_006246539.1 | 6 | 571 | |||
XM_006246478.2 | XP_006246540.1 | 5 | 581 | |||
XM_006246479.2 | XP_006246541.1 | 4 | 379 | |||
XM_006246483.1 | XP_006246545.1 | 6 | 486 | |||
XM_008767812.1 | XP_008766034.1 | 6 | 665 | |||
XM_008767813.1 | XP_008766035.1 | 5 | 1240 | |||
XM_008767814.1 | XP_008766036.1 | 5 | 489 | |||
XM_008767815.1 | XP_008766037.1 | 5 | 1236 | |||
GenBank mRNA | AF354443.1 | - | 5 | 385 | ||
FM065046.1 | - | 5 | 434 |
