siRNA Details
Gene Symbol: Wnt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107005.2 | NP_001100475.2 | 4 | 918 | ||
XM_006246487.2 | XP_006246549.1 | 4 | 1076 |

More Information
Other Species Targeted

Gene Ontology Categories:
