siRNA Details
Gene Symbol: Rai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271207.1 | NP_001258136.1 | 2 | 1877 | ||
XM_008767818.1 | XP_008766040.1 | 1 | 1591 | |||
XM_008767819.1 | XP_008766041.1 | 2 | 2521 |
