siRNA Details
Gene Symbol: Slc46a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013969.1 | NP_001013991.1 | 2 | 437 | ||
XM_006246939.2 | XP_006247001.1 | 3 | 745 | |||
GenBank mRNA | BC089868.1 | - | 2 | 437 | 7322786 | |
FQ231269.1 | - | 2 | 463 |
