siRNA Details
Gene Symbol: Rasl10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191648.1 | NP_001178577.1 | 2 | 257 | ||
XM_006247028.2 | XP_006247090.1 | 2 | 723 | |||
XM_006247029.2 | XP_006247091.1 | 3 | 943 | |||
XM_006247030.2 | XP_006247092.1 | 2 | 940 | |||
XM_008768033.1 | XP_008766255.1 | 2 | 2139 | |||
XM_008768034.1 | XP_008766256.1 | 2 | 723 |
Gene Symbol: LOC103694863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773805.1 | XP_008772027.1 | 2 | 2139 | ||
XM_008773806.1 | XP_008772028.1 | 2 | 723 | |||
XM_008773807.1 | XP_008772029.1 | 3 | 943 | |||
XM_008773808.1 | XP_008772030.1 | 2 | 940 | |||
XM_008773809.1 | XP_008772031.1 | 2 | 723 | |||
XM_008773810.1 | XP_008772032.1 | 2 | 723 |
