siRNA Details
Gene Symbol: Dlx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107040.1 | NP_001100510.1 | 3 | 548 | ||
GenBank mRNA | HQ616894.1 | - | 3 | 570 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dlx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107040.1 | NP_001100510.1 | 3 | 548 | ||
GenBank mRNA | HQ616894.1 | - | 3 | 570 |