siRNA Details
Gene Symbol: Krt16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008752.1 | NP_001008752.1 | 3 | 617 | ||
XM_006247309.2 | XP_006247371.1 | 3 | 695 |
Gene Symbol: Krt14l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | D63774.1 | - | 407 |
