siRNA Details
Gene Symbol: Rundc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198758.2 | NP_942053.2 | 3 | 479 | ||
XM_006247335.2 | XP_006247397.1 | 3 | 501 | |||
XM_006247337.2 | XP_006247399.1 | 3 | 501 | |||
XM_006247339.2 | XP_006247401.1 | 3 | 501 | |||
XM_008768059.1 | XP_008766281.1 | 3 | 501 | |||
GenBank mRNA | CB797318.1 | - | 2 | 160 | ||
DY319760.1 | - | 3 | 513 |
