siRNA Details
Gene Symbol: Rundc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198758.2 | NP_942053.2 | 6 | 824 | ||
XM_006247335.2 | XP_006247397.1 | 6 | 846 | |||
XM_006247337.2 | XP_006247399.1 | 6 | 846 | |||
XM_006247339.2 | XP_006247401.1 | 6 | 846 | |||
XM_006247340.2 | XP_006247402.1 | 6 | 831 | |||
XM_008768056.1 | XP_008766278.1 | 6 | 834 | |||
XM_008768058.1 | XP_008766280.1 | 6 | 831 | |||
XM_008768059.1 | XP_008766281.1 | 6 | 846 | |||
GenBank mRNA | BC127472.1 | - | 5 | 408 | ||
BC158572.1 | - | 3 | 221 |
