siRNA Details
Gene Symbol: Efcab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013972.2 | NP_001013994.1 | 3, 4 | 458 | ||
GenBank mRNA | BC081930.1 | - | 3, 4 | 347 | ||
BC127522.1 | - | 3, 4 | 458 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Efcab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013972.2 | NP_001013994.1 | 3, 4 | 458 | ||
GenBank mRNA | BC081930.1 | - | 3, 4 | 347 | ||
BC127522.1 | - | 3, 4 | 458 |