siRNA Details
Gene Symbol: Tex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247645.1 | XP_006247707.1 | 4 | 2136 | ||
XM_006247646.2 | XP_006247708.1 | 4 | 2968 | |||
XM_006247647.1 | XP_006247709.1 | 4 | 2136 | |||
GenBank mRNA | BC083806.1 | - | 3 | 520 |
